Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047434.2 | NP_001040899.1 | 1 | 170 | ||
NM_206831.2 | NP_996662.1 | 1 | 170 | |||
GenBank mRNA | AK022970.1 | - | 1 | 150 | ||
AK092040.1 | - | 1 | 123 | |||
BC010181.2 | - | 1 | 79 | 4543414 | ||
DA447430.1 | - | 1 | 160 | |||
DA875039.1 | - | 1 | 170 |