Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131034.3 | NP_001124506.1 | 3 | 283 | ||
NM_001193318.2 | NP_001180247.1 | 3 | 283 | |||
NM_194439.4 | NP_919420.1 | 3 | 283 | |||
XM_005272274.2 | XP_005272331.1 | 3 | 281 | |||
XM_011513439.1 | XP_011511741.1 | 4 | 472 | |||
XM_011513443.1 | XP_011511745.1 | 2 | 451 | |||
XM_011513444.1 | XP_011511746.1 | 3 | 281 | |||
XM_011513445.2 | XP_011511747.1 | 3 | 295 | |||
XM_011513446.1 | XP_011511748.1 | 3 | 287 | |||
XM_017008039.1 | XP_016863528.1 | 3 | 309 | |||
XM_017008040.1 | XP_016863529.1 | 4 | 347 | |||
XM_017008041.1 | XP_016863530.1 | 3 | 299 | |||
XM_017008042.1 | XP_016863531.1 | 3 | 285 | |||
GenBank mRNA | AK096160.1 | - | 3 | 281 | ||
AK310043.1 | - | 4 | 472 | |||
BC036250.1 | - | 3 | 249 | 5267536 | ||
BC050356.1 | - | 3 | 279 | 4815093 | ||
BC101259.1 | - | 3 | 245 | 40023273 | ||
BC101260.1 | - | 3 | 245 | 40023274 | ||
BG701603.1 | - | 3 | 284 | |||
BI460105.1 | - | 3 | 254 |