Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RELL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130029.1 | NP_001123501.1 | 3, 4 | 1109 | ||
NM_173828.4 | NP_776189.3 | 2, 3 | 1537 | |||
XM_005268414.3 | XP_005268471.1 | 3, 4 | 1113 | |||
XM_011537624.1 | XP_011535926.1 | 3, 4 | 1129 | |||
XM_011537625.2 | XP_011535927.1 | 2, 3 | 1541 | |||
GenBank mRNA | AK075064.1 | - | 2, 3 | 364 | ||
AY358163.1 | - | 2, 3 | 1473 | |||
BC063469.1 | - | 3, 4 | 578 | 5211730 |