Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175921.5 | NP_787117.3 | 4 | 436 | ||
XM_005248289.4 | XP_005248346.1 | 4 | 467 | |||
XM_011514032.2 | XP_011512334.1 | 5 | 565 | |||
GenBank mRNA | BF439305.1 | - | 4 | 400 | ||
HY116483.1 | - | 4 | 437 |