Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173685.2 | NP_775956.1 | 3, 4 | 376 | ||
XM_005250875.2 | XP_005250932.1 | 3, 4 | 384 | |||
XM_005250876.4 | XP_005250933.1 | 2, 3 | 277 | |||
XM_011516974.2 | XP_011515276.1 | 4, 5 | 446 | |||
XM_011516975.1 | XP_011515277.1 | 3, 4 | 341 | |||
XM_017013331.1 | XP_016868820.1 | 4, 5 | 414 | |||
GenBank mRNA | AK057002.1 | - | 3, 4 | 346 | ||
BC032797.1 | - | 3, 4 | 375 | 5109691 |