Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173685.2 | NP_775956.1 | 4, 5 | 500 | ||
XM_005250875.2 | XP_005250932.1 | 4, 5 | 508 | |||
XM_005250876.4 | XP_005250933.1 | 3, 4 | 401 | |||
XM_011516974.2 | XP_011515276.1 | 5, 6 | 570 | |||
XM_011516975.1 | XP_011515277.1 | 4, 5 | 465 | |||
XM_017013330.1 | XP_016868819.1 | 2, 3 | 681 | |||
XM_017013331.1 | XP_016868820.1 | 5, 6 | 538 | |||
XM_017013332.1 | XP_016868821.1 | 2, 3 | 699 | |||
XM_017013333.1 | XP_016868822.1 | 5, 6 | 610 | |||
XM_017013334.1 | XP_016868823.1 | 2, 3 | 721 | |||
GenBank mRNA | AK057002.1 | - | 4, 5 | 470 | ||
BC032797.1 | - | 4, 5 | 499 | 5109691 |