Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181723.2 | NP_859074.1 | 11 | 1180 | ||
XM_005273477.2 | XP_005273534.1 | 10 | 1139 | |||
XM_006716328.2 | XP_006716391.1 | 10 | 1082 | |||
XM_006716329.2 | XP_006716392.1 | 9 | 1043 | |||
XM_011544507.2 | XP_011542809.1 | 11 | 1160 | |||
XM_017013336.1 | XP_016868825.1 | 10 | 1121 | |||
XM_017013340.1 | XP_016868829.1 | 10 | 1125 | |||
XM_017013342.1 | XP_016868831.1 | 9 | 992 | |||
GenBank mRNA | AL833419.1 | - | 1 | 64 | ||
BC032868.1 | - | 11 | 1135 | |||
BC045553.1 | - | 11 | 1179 | 5262603 |