Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181723.2 | NP_859074.1 | 2 | 492 | ||
XM_005273477.2 | XP_005273534.1 | 2 | 490 | |||
XM_006716328.2 | XP_006716391.1 | 2 | 490 | |||
XM_006716329.2 | XP_006716392.1 | 2 | 490 | |||
XM_006716330.2 | XP_006716393.1 | 2 | 490 | |||
XM_006716333.3 | XP_006716396.1 | 2 | 490 | |||
XM_011544507.2 | XP_011542809.1 | 2 | 490 | |||
XM_011544508.2 | XP_011542810.1 | 2 | 490 | |||
XM_017013336.1 | XP_016868825.1 | 2 | 490 | |||
XM_017013337.1 | XP_016868826.1 | 2 | 490 | |||
XM_017013338.1 | XP_016868827.1 | 2 | 490 | |||
XM_017013339.1 | XP_016868828.1 | 2 | 490 | |||
XM_017013340.1 | XP_016868829.1 | 2 | 469 | |||
XM_017013341.1 | XP_016868830.1 | 2 | 490 | |||
XM_017013342.1 | XP_016868831.1 | 2 | 375 | |||
XM_017013343.1 | XP_016868832.1 | 2 | 490 | |||
GenBank mRNA | BC032868.1 | - | 2 | 447 | ||
BC045553.1 | - | 2 | 491 | 5262603 | ||
BI546243.1 | - | 3 | 352 |