Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173687.3 | NP_775958.1 | 5 | 788 | ||
NR_126398.1 | 7 | 970 | ||||
NR_126399.1 | 5 | 779 | ||||
GenBank mRNA | AK094450.1 | - | 5 | 736 | ||
BC063497.1 | - | 5 | 690 | |||
BC073830.1 | - | 7 | 918 | |||
BC120993.2 | - | 5 | 714 |