Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271156.2 | NP_001258085.2 | 2 | 1551 | ||
NM_173832.5 | NP_776193.2 | 2 | 837 | |||
GenBank mRNA | AK096024.1 | - | 2 | 826 | ||
AL831951.1 | - | 2 | 1262 | |||
BC034608.1 | - | 2 | 1551 | 4827717 | ||
CR992458.1 | - | 1 | 283 | |||
DA216172.1 | - | 1 | 481 |