Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304533.1 | NP_001291462.1 | 2 | 300 | ||
NR_130764.1 | 2 | 300 | ||||
XM_011517511.2 | XP_011515813.1 | 2 | 300 | |||
XM_011517512.2 | XP_011515814.1 | 2 | 300 | |||
XM_017013359.1 | XP_016868848.1 | 2 | 300 | |||
XM_017013360.1 | XP_016868849.1 | 2 | 300 | |||
XM_017013361.1 | XP_016868850.1 | 2 | 1682 | |||
GenBank mRNA | AK096949.1 | - | 2 | 300 | ||
AK299689.1 | - | 2 | 573 |