Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LURAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203403.1 | NP_981948.1 | 2 | 1051 | ||
GenBank mRNA | AB073606.1 | - | 2 | 1053 | ||
AK074752.1 | - | 2 | 532 | |||
AK095824.1 | - | 2 | 1050 | |||
BC036923.1 | - | 2 | 1026 | 5228515 | ||
BM739457.1 | - | 2 | 267 |