Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152467.3 | NP_689680.2 | 5 | 1768 | ||
XM_011524706.2 | XP_011523008.1 | 7 | 1901 | |||
GenBank mRNA | AK057224.1 | - | 5 | 1731 | ||
AK301797.1 | - | 5 | 1672 | |||
AY495339.1 | - | 5 | 1750 | |||
BC067753.1 | - | 5 | 1755 |