Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042518.1 | NP_001035983.1 | 1, 2 | 191 | ||
NM_175739.3 | NP_783866.2 | 1, 2 | 191 | |||
GenBank mRNA | AK301082.1 | - | 1, 2 | 85 | ||
AY185496.1 | - | 1, 2 | 191 | |||
AY220118.1 | - | 1, 2 | 105 | |||
AY220119.1 | - | 1, 2 | 69 | |||
AY358700.1 | - | 1, 2 | 35 | |||
BC111498.1 | - | 1, 2 | 94 | 5498091 |