Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042518.1 | NP_001035983.1 | 4 | 561 | ||
NM_001284275.1 | NP_001271204.1 | 3 | 671 | |||
NM_001284276.1 | NP_001271205.1 | 4 | 568 | |||
NM_175739.3 | NP_783866.2 | 3 | 861 | |||
XM_011536714.2 | XP_011535016.1 | 3 | 967 | |||
XM_011536715.2 | XP_011535017.1 | 4 | 831 | |||
XM_011536716.2 | XP_011535018.1 | 3 | 777 | |||
XM_011536717.1 | XP_011535019.1 | 3 | 1660 | |||
GenBank mRNA | AK301082.1 | - | 3 | 755 | ||
AY185496.1 | - | 3 | 861 | |||
AY185497.1 | - | 3 | 671 | |||
BC111498.1 | - | 3 | 764 | 5498091 |