Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175856.4 | NP_787052.3 | 3 | 2288 | ||
XM_011543363.2 | XP_011541665.1 | 3 | 2141 | |||
XM_011543364.2 | XP_011541666.1 | 2 | 1497 | |||
XM_017009434.1 | XP_016864923.1 | 2 | 11187 | |||
GenBank mRNA | AB086062.1 | - | 3 | 2288 | ||
AB175496.1 | - | 3 | 2288 | |||
AJ504664.2 | - | 3 | 1825 | |||
AJ578034.2 | - | 3 | 2646 | |||
AY726604.1 | - | 3 | 1590 | |||
BC137325.1 | - | 3 | 2357 | 9021322 |