Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP11-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175858.2 | NP_787054.1 | 1 | 418 | ||
GenBank mRNA | AB096963.1 | - | 1 | 396 | ||
AJ457065.1 | - | 1 | 418 | |||
BC130555.1 | - | 1 | 411 | 40146586 | ||
BC130557.1 | - | 1 | 411 | 40146588 |