Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176822.3 | NP_789792.1 | 10 | 3268 | ||
XM_011520044.1 | XP_011518346.1 | 9 | 3120 | |||
GenBank mRNA | AK310900.1 | - | 10 | 3268 | ||
AY154469.1 | - | 9 | 2945 |