Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004470.1 | NP_001004470.1 | 6 | 547 | ||
XM_017016174.1 | XP_016871663.1 | 5 | 539 | |||
GenBank mRNA | AB062101.1 | - | 6 | 547 | ||
AJ621583.1 | - | 6 | 547 | |||
BC137102.1 | - | 6 | 679 | 9021099 | ||
BC137103.1 | - | 6 | 679 | 9021100 |