Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300972.1 | NP_001287901.1 | 5 | 1421 | ||
NM_001300973.1 | NP_001287902.1 | 5 | 1418 | |||
NM_001300975.1 | NP_001287904.1 | 5 | 1421 | |||
NM_001300976.1 | NP_001287905.1 | 5 | 1421 | |||
NM_001300977.1 | NP_001287906.1 | 5 | 1421 | |||
NM_182603.3 | NP_872409.2 | 5 | 1337 | |||
NR_125355.1 | 5 | 727 | ||||
GenBank mRNA | AK095193.1 | - | 5 | 567 | ||
AK310765.1 | - | 5 | 715 | |||
BC045621.1 | - | 5 | 1421 | 4813915 | ||
BC171752.1 | - | 5 | 515 | |||
BX648690.1 | - | 5 | 911 | |||
DQ508815.1 | - | 5 | 762 | |||
DQ508934.1 | - | 5 | 1448 | |||
EF041819.1 | - | 5 | 1281 |