Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178539.4 | NP_848634.1 | 2 | 1666 | ||
XM_006719370.2 | XP_006719433.1 | 3 | 1395 | |||
XM_011538272.1 | XP_011536574.1 | 3 | 1728 | |||
XM_011538274.1 | XP_011536576.1 | 3 | 1119 | |||
GenBank mRNA | AK123580.1 | - | 3 | 274 | ||
AL834160.1 | - | 2 | 1018 | |||
AY325115.1 | - | 2 | 239 | |||
BC028403.1 | - | 2 | 615 | 4818605 | ||
BC050347.1 | - | 2 | 1597 | |||
CR749367.1 | - | 5 | 574 |