Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM19A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178539.4 | NP_848634.1 | 4 | 1874 | ||
XM_006719370.2 | XP_006719433.1 | 5 | 1603 | |||
XM_011538272.1 | XP_011536574.1 | 5 | 1936 | |||
XM_011538274.1 | XP_011536576.1 | 5 | 1327 | |||
GenBank mRNA | AK123580.1 | - | 5 | 482 | ||
AL834160.1 | - | 4 | 1226 | |||
AY325115.1 | - | 4 | 447 | |||
BC028403.1 | - | 4 | 823 | 4818605 | ||
BC040286.1 | - | 3 | 266 | |||
BC050347.1 | - | 4 | 1805 | |||
CR749367.1 | - | 7 | 782 |