Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005920.2 | NP_001005920.2 | 8 | 711 | ||
NM_001323918.1 | NP_001310847.1 | 8 | 718 | |||
NM_001323920.1 | NP_001310849.1 | 7 | 628 | |||
NM_001323922.1 | NP_001310851.1 | 7 | 628 | |||
NR_136650.1 | 7 | 806 | ||||
NR_136651.1 | 7 | 806 | ||||
NR_136652.1 | 6 | 716 | ||||
GenBank mRNA | AF370420.1 | - | 8 | 595 | ||
AL524336.3 | - | 7 | 691 | |||
AL524922.3 | - | 7 | 693 | |||
BC073785.1 | - | 7 | 683 | |||
BC094850.1 | - | 5 | 329 | |||
BC137100.1 | - | 8 | 677 | 9021097 | ||
BC137101.1 | - | 8 | 677 | 9021098 | ||
BG284477.1 | - | 3 | 168 | |||
BG423721.1 | - | 4 | 228 | |||
BG826580.1 | - | 5 | 509 | |||
BI767435.1 | - | 7 | 531 | |||
BX335015.2 | - | 8 | 597 | |||
DQ074695.1 | - | 7 | 501 |