Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM92B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198491.2 | NP_940893.1 | 7 | 746 | ||
XM_005255931.2 | XP_005255988.1 | 7 | 746 | |||
XM_011523063.1 | XP_011521365.1 | 7 | 746 | |||
XM_017023198.1 | XP_016878687.1 | 7 | 746 | |||
GenBank mRNA | AK126284.1 | - | 7 | 746 | ||
BC093665.1 | - | 7 | 628 | 7939510 | ||
BC111944.1 | - | 7 | 628 | 8327412 |