Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP27C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001665.3 | NP_001001665.3 | 8 | 1226 | ||
XM_017003960.1 | XP_016859449.1 | 9 | 1416 | |||
GenBank mRNA | AK131190.1 | - | 8 | 1207 | ||
BC039307.1 | - | 8 | 1226 | 5266370 |