Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308370.1 | NP_001295299.1 | 4 | 1803 | ||
NM_194312.3 | NP_919288.2 | 9 | 2876 | |||
XM_011511087.1 | XP_011509389.1 | 2 | 1647 | |||
GenBank mRNA | AK124559.1 | - | 9 | 2801 | ||
AK124612.1 | - | 6 | 1943 | |||
AK128187.1 | - | 2 | 1647 | |||
BC042051.1 | - | 5 | 1867 | |||
BC080639.1 | - | 4 | 1803 | 6597009 |