Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178554.4 | NP_848649.3 | 10 | 1071 | ||
XM_005247418.3 | XP_005247475.1 | 9 | 6039 | |||
XM_006713612.3 | XP_006713675.1 | 9 | 5985 | |||
XM_017006287.1 | XP_016861776.1 | 10 | 6117 | |||
XM_017006288.1 | XP_016861777.1 | 10 | 6063 | |||
XM_017006289.1 | XP_016861778.1 | 9 | 6054 | |||
XM_017006290.1 | XP_016861779.1 | 9 | 6000 | |||
GenBank mRNA | AK090526.1 | - | 10 | 1071 | ||
AK126993.1 | - | 9 | 846 | |||
AK293840.1 | - | 9 | 1004 | |||
DA212239.1 | - | 2 | 281 |