Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042784.1 | NP_001036249.1 | 4, 5 | 542 | ||
XM_005262974.4 | XP_005263031.1 | 5, 6 | 669 | |||
XM_006714204.3 | XP_006714267.1 | 5, 6 | 669 | |||
XM_011531908.2 | XP_011530210.1 | 6, 7 | 773 | |||
XM_011531909.2 | XP_011530211.1 | 6, 7 | 789 | |||
XM_011531910.2 | XP_011530212.1 | 5, 6 | 1250 | |||
XM_011531911.1 | XP_011530213.1 | 5, 6 | 633 | |||
XM_011531912.2 | XP_011530214.1 | 5, 6 | 529 | |||
XM_011531913.1 | XP_011530215.1 | 4, 5 | 557 | |||
XM_011531914.2 | XP_011530216.1 | 4, 5 | 626 | |||
XM_011531915.2 | XP_011530217.1 | 5, 6 | 670 | |||
XM_011531916.2 | XP_011530218.1 | 4, 5 | 451 | |||
XM_011531917.1 | XP_011530219.1 | 5, 6 | 494 | |||
XM_017008160.1 | XP_016863649.1 | 6, 7 | 1283 | |||
GenBank mRNA | AK098636.1 | - | 3, 4 | 349 |