Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181789.2 | NP_861454.2 | 10 | 1774 | ||
XM_011521501.2 | XP_011519803.1 | 10 | 3631 | |||
XM_017022121.1 | XP_016877610.1 | 9 | 1617 | |||
XM_017022122.1 | XP_016877611.1 | 10 | 1805 | |||
XM_017022123.1 | XP_016877612.1 | 10 | 1596 | |||
XM_017022124.1 | XP_016877613.1 | 10 | 1648 | |||
GenBank mRNA | AK172756.1 | - | 10 | 1610 | ||
AY358144.1 | - | 9 | 1235 | |||
BC113397.1 | - | 10 | 1260 | 8322449 | ||
BC113399.1 | - | 10 | 1260 | 8322451 | ||
BX538105.1 | - | 10 | 1283 |