Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114974.1 | NP_001108446.1 | 4 | 855 | ||
NM_198501.2 | NP_940903.2 | 4 | 507 | |||
XM_005256622.3 | XP_005256679.1 | 3 | 684 | |||
XM_006721515.2 | XP_006721578.1 | 4 | 1023 | |||
GenBank mRNA | AK124452.1 | - | 4 | 489 | ||
BC105050.1 | - | 4 | 408 | 8144053 | ||
BC110859.1 | - | 4 | 855 | 5190568 | ||
BC112177.1 | - | 4 | 408 | 8327645 | ||
DA822419.1 | - | 4 | 507 |