Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMTNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114974.1 | NP_001108446.1 | 7 | 1307 | ||
NM_198501.2 | NP_940903.2 | 7 | 959 | |||
XM_005256622.3 | XP_005256679.1 | 6 | 1136 | |||
XM_006721515.2 | XP_006721578.1 | 7 | 1475 | |||
GenBank mRNA | AK124452.1 | - | 7 | 941 | ||
BC105050.1 | - | 7 | 860 | 8144053 | ||
BC110859.1 | - | 7 | 1307 | 5190568 | ||
BC112177.1 | - | 7 | 860 | 8327645 |