Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287819.1 | NP_001274748.1 | 21 | 2584 | ||
NM_001287820.1 | NP_001274749.1 | 20 | 2434 | |||
NM_198503.3 | NP_940905.2 | 22 | 2656 | |||
XM_006711294.2 | XP_006711357.1 | 22 | 3130 | |||
XM_006711295.2 | XP_006711358.1 | 21 | 3058 | |||
XM_011509482.2 | XP_011507784.1 | 21 | 3055 | |||
XM_017001178.1 | XP_016856667.1 | 21 | 3058 | |||
XM_017001179.1 | XP_016856668.1 | 22 | 3130 | |||
XM_017001181.1 | XP_016856670.1 | 12 | 1645 | |||
XM_017001182.1 | XP_016856671.1 | 21 | 3021 | |||
XM_017001183.1 | XP_016856672.1 | 9 | 1181 | |||
XM_017001184.1 | XP_016856673.1 | 21 | 3021 | |||
XM_017001185.1 | XP_016856674.1 | 20 | 2983 | |||
GenBank mRNA | AK127807.1 | - | 14 | 1857 | ||
AK131498.1 | - | 20 | 2659 | |||
AK309729.1 | - | 21 | 2573 | |||
AY359444.1 | - | 22 | 2594 | |||
BC103948.1 | - | 21 | 2512 | 40009146 | ||
BC103949.1 | - | 20 | 2362 | 40009147 | ||
BC103950.1 | - | 21 | 2512 | 40009148 | ||
BX647852.1 | - | 21 | 2566 | |||
EU251076.1 | - | 22 | 2534 |