Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287819.1 | NP_001274748.1 | 24 | 2908 | ||
NM_001287820.1 | NP_001274749.1 | 23 | 2758 | |||
NM_198503.3 | NP_940905.2 | 25 | 2980 | |||
XM_006711294.2 | XP_006711357.1 | 25 | 3454 | |||
XM_006711295.2 | XP_006711358.1 | 24 | 3382 | |||
XM_011509482.2 | XP_011507784.1 | 24 | 3379 | |||
XM_017001178.1 | XP_016856667.1 | 24 | 3382 | |||
XM_017001181.1 | XP_016856670.1 | 15 | 1969 | |||
XM_017001182.1 | XP_016856671.1 | 24 | 3345 | |||
XM_017001183.1 | XP_016856672.1 | 12 | 1505 | |||
XM_017001184.1 | XP_016856673.1 | 24 | 3345 | |||
XM_017001185.1 | XP_016856674.1 | 23 | 3307 | |||
GenBank mRNA | AK127807.1 | - | 17 | 2181 | ||
AK131498.1 | - | 23 | 2983 | |||
AK309729.1 | - | 24 | 2897 | |||
AY359444.1 | - | 25 | 2918 | |||
BC103947.1 | - | 19 | 2172 | |||
BC103948.1 | - | 24 | 2836 | 40009146 | ||
BC103949.1 | - | 23 | 2686 | 40009147 | ||
BC103950.1 | - | 24 | 2836 | 40009148 | ||
BX647852.1 | - | 24 | 2890 | |||
EU251076.1 | - | 25 | 2858 |