Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304366.1 | NP_001291295.1 | 9 | 1584 | ||
NM_182610.3 | NP_872416.1 | 9 | 1607 | |||
NR_130713.1 | 10 | 1697 | ||||
GenBank mRNA | BC117339.1 | - | 8 | 1291 | 40125890 | |
BC143909.1 | - | 9 | 1403 | |||
BX537903.1 | - | 9 | 1487 | |||
BX648534.1 | - | 9 | 1465 | |||
BX648989.1 | - | 9 | 1487 | |||
BX648991.1 | - | 9 | 1585 |