Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304366.1 | NP_001291295.1 | 5, 6 | 666 | ||
NM_182610.3 | NP_872416.1 | 5, 6 | 689 | |||
NR_130713.1 | 6, 7 | 779 | ||||
GenBank mRNA | BC117339.1 | - | 4, 5 | 373 | 40125890 | |
BC143909.1 | - | 5, 6 | 485 | |||
BX537903.1 | - | 5, 6 | 569 | |||
BX648534.1 | - | 5, 6 | 547 | |||
BX648989.1 | - | 5, 6 | 569 | |||
BX648991.1 | - | 5, 6 | 667 |