Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AADACL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207365.3 | NP_997248.2 | 5 | 1120 | ||
XM_011512753.1 | XP_011511055.1 | 5 | 1148 | |||
GenBank mRNA | BC065724.1 | - | 5 | 1113 | 6663150 | |
BX118873.1 | - | 1 | 327 | |||
BX647585.1 | - | 4 | 897 |