Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198504.2 | NP_940906.1 | 1 | 777 | ||
GenBank mRNA | AK123932.1 | - | 1 | 777 | ||
AY424287.1 | - | 1 | 854 | |||
BC118666.1 | - | 1 | 777 | 40016554 | ||
BC122527.1 | - | 1 | 777 | 40016555 |
Set Membership: |
G-protein coupled receptor |