Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183375.2 | NP_899231.2 | 4 | 599 | ||
XM_011531921.2 | XP_011530223.1 | 3 | 1045 | |||
XM_011531922.2 | XP_011530224.1 | 2 | 861 | |||
GenBank mRNA | BC121109.1 | - | 4 | 603 | 40120369 | |
BC121110.1 | - | 2 | 174 | 40120371 |