Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182762.3 | NP_877439.3 | 5 | 1891 | ||
GenBank mRNA | AJ313524.1 | - | 2 | 1582 | ||
AK128850.1 | - | 1 | 1052 | |||
AK131400.1 | - | 5 | 1891 | |||
BC137090.1 | - | 3 | 1612 | 9021087 |