Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178839.4 | NP_849161.2 | 2 | 836 | ||
XM_017003986.1 | XP_016859475.1 | 2 | 1368 | |||
XM_017003987.1 | XP_016859476.1 | 2 | 1216 | |||
GenBank mRNA | AK056644.1 | - | 2 | 867 | ||
AK225779.1 | - | 2 | 1223 | |||
AK290512.1 | - | 2 | 1086 | |||
AY182024.1 | - | 1 | 566 | |||
AY358310.1 | - | 2 | 1285 | |||
BC045113.1 | - | 2 | 836 | 5200499 |