Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178130.3 | NP_835231.1 | 8 | 538 | ||
XM_005247429.3 | XP_005247486.1 | 8 | 519 | |||
XM_006713615.3 | XP_006713678.1 | 7 | 2401 | |||
XM_011512774.2 | XP_011511076.1 | 7 | 405 | |||
XM_011512777.2 | XP_011511079.1 | 7 | 405 | |||
XM_017006306.1 | XP_016861795.1 | 8 | 522 | |||
XM_017006307.1 | XP_016861796.1 | 7 | 971 | |||
XM_017006308.1 | XP_016861797.1 | 8 | 2518 | |||
XM_017006309.1 | XP_016861798.1 | 8 | 550 | |||
XM_017006310.1 | XP_016861799.1 | 7 | 480 | |||
XM_017006311.1 | XP_016861800.1 | 7 | 405 | |||
XM_017006312.1 | XP_016861801.1 | 7 | 2401 | |||
XM_017006313.1 | XP_016861802.1 | 9 | 3025 | |||
XM_017006314.1 | XP_016861803.1 | 8 | 976 | |||
XM_017006315.1 | XP_016861804.1 | 6 | 454 | |||
XM_017006316.1 | XP_016861805.1 | 6 | 2329 | |||
XM_017006317.1 | XP_016861806.1 | 6 | 454 | |||
XM_017006318.1 | XP_016861807.1 | 5 | 782 | |||
XM_017006319.1 | XP_016861808.1 | 3 | 189 | |||
GenBank mRNA | AF196568.1 | - | 7 | 511 | ||
AK094830.1 | - | 8 | 538 | |||
AY336746.1 | - | 4 | 228 | |||
BC107054.2 | - | 5 | 332 | 40005048 |