Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMIGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143668.1 | NP_001137140.1 | 3 | 1094 | ||
NM_181847.4 | NP_862830.1 | 2 | 992 | |||
XM_005268837.3 | XP_005268894.1 | 3 | 1974 | |||
GenBank mRNA | AB080610.1 | - | 1 | 502 | ||
AK091761.1 | - | 2 | 815 | |||
AL833007.1 | - | 3 | 790 | |||
AY237005.1 | - | 1 | 439 | |||
AY454159.1 | - | 2 | 907 | |||
BC047595.1 | - | 2 | 907 | 4828334 | ||
BC095477.1 | - | 3 | 1094 | 30387316 |