Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194314.2 | NP_919290.2 | 1 | 970 | ||
NR_135153.1 | 1 | 970 | ||||
XM_011509507.2 | XP_011507809.1 | 1 | 949 | |||
XM_017001205.1 | XP_016856694.1 | 1 | 949 | |||
GenBank mRNA | AK128549.1 | - | 1 | 949 | ||
AY163816.2 | - | 1 | 970 | |||
BC132818.1 | - | 1 | 934 | 40146840 | ||
BC136600.1 | - | 1 | 934 | 9020590 |