Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198526.3 | NP_940928.2 | 4 | 1898 | ||
XM_005254906.3 | XP_005254963.1 | 4 | 1924 | |||
XM_005254907.3 | XP_005254964.1 | 4 | 1845 | |||
XM_011521540.2 | XP_011519842.1 | 5 | 2006 | |||
XM_011521541.2 | XP_011519843.1 | 5 | 1967 | |||
XM_011521542.2 | XP_011519844.1 | 4 | 1885 | |||
XM_011521543.2 | XP_011519845.1 | 3 | 1774 | |||
XM_011521544.1 | XP_011519846.1 | 3 | 1774 | |||
GenBank mRNA | AK094712.1 | - | 4 | 1869 | ||
AK160373.1 | - | 3 | 588 | |||
AL831826.1 | - | 3 | 369 | |||
BC126476.1 | - | 3 | 1770 | 8992192 | ||
BC130541.1 | - | 3 | 1770 | 40146572 |