Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 10, 11 | 1332 | ||
NM_001030312.2 | NP_001025483.1 | 6, 7 | 915 | |||
NM_001030313.2 | NP_001025484.1 | 7, 8 | 1047 | |||
NM_001160277.1 | NP_001153749.1 | 9, 10 | 1200 | |||
NM_201548.4 | NP_963842.1 | 9, 10 | 1254 | |||
NR_027689.1 | 7, 8 | 1058 | ||||
NR_027690.1 | 8, 9 | 1190 | ||||
GenBank mRNA | AJ640141.1 | - | 9, 10 | 1171 | ||
AJ697856.1 | - | 8, 9 | 1089 | |||
AK129976.1 | - | 6, 7 | 497 | |||
AK293844.1 | - | 9, 10 | 1201 | |||
AY357073.1 | - | 9, 10 | 1153 | |||
AY690329.1 | - | 10, 11 | 1231 | |||
AY690330.1 | - | 6, 7 | 814 | |||
AY690331.1 | - | 7, 8 | 946 | |||
AY690332.1 | - | 7, 8 | 957 | |||
AY690333.1 | - | 8, 9 | 1089 | |||
BC137498.1 | - | 9, 10 | 1180 | 9021498 | ||
BC137499.1 | - | 9, 10 | 1180 | 9021499 | ||
HQ426666.1 | - | 9, 10 | 1060 | |||
HQ426667.1 | - | 8, 9 | 996 | |||
HQ426668.1 | - | 7, 8 | 864 | |||
HQ426669.1 | - | 7, 8 | 853 | |||
HQ426670.1 | - | 6, 7 | 721 | |||
HQ426671.1 | - | 10, 11 | 1060 | |||
HQ426672.1 | - | 9, 10 | 982 | |||
HQ426673.1 | - | 8, 9 | 918 | |||
HQ426674.1 | - | 7, 8 | 786 | |||
HQ426675.1 | - | 7, 8 | 775 | |||
HQ426676.1 | - | 6, 7 | 643 | |||
HQ426677.1 | - | 7, 8 | 590 | |||
HQ426678.1 | - | 10, 11 | 1307 | |||
HQ426679.1 | - | 7, 8 | 1612 | |||
JF290420.1 | - | 9, 10 | 1140 |