Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320865.1 | NP_001307794.1 | 6 | 678 | ||
NM_198559.1 | NP_940961.1 | 6 | 645 | |||
XM_005246539.4 | XP_005246596.1 | 5 | 642 | |||
XM_005246541.4 | XP_005246598.1 | 4 | 441 | |||
XM_011511149.2 | XP_011509451.1 | 4 | 440 | |||
XM_011511150.2 | XP_011509452.1 | 4 | 406 | |||
XM_017004053.1 | XP_016859542.1 | 4 | 841 | |||
GenBank mRNA | BC052750.1 | - | 6 | 645 | 5742766 |