Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167962.1 | NP_001161434.1 | 8, 9 | 1149 | ||
NM_001321787.1 | NP_001308716.1 | 8, 9 | 1149 | |||
NM_198999.2 | NP_945350.1 | 8, 9 | 1149 | |||
NM_206883.2 | NP_996766.1 | 8, 9 | 1149 | |||
NM_206884.2 | NP_996767.1 | 8, 9 | 1149 | |||
NM_206885.2 | NP_996768.1 | 8, 9 | 1149 | |||
NR_120441.1 | 7, 8 | 1069 | ||||
NR_120442.1 | 7, 8 | 1017 | ||||
NR_120443.1 | 7, 8 | 1069 | ||||
NR_135801.1 | 8, 9 | 1149 | ||||
NR_135802.1 | 8, 9 | 1149 | ||||
XM_011516170.2 | XP_011514472.1 | 8, 9 | 1939 | |||
GenBank mRNA | AF523354.1 | - | 8, 9 | 1123 | ||
AK126898.1 | - | 7, 8 | 986 | |||
AY256823.1 | - | 8, 9 | 1123 | |||
AY256824.1 | - | 8, 9 | 1123 | |||
AY256825.1 | - | 8, 9 | 1123 | |||
AY289133.1 | - | 6, 7 | 895 | |||
AY289134.1 | - | 6, 7 | 895 | |||
BC100832.1 | - | 7, 8 | 1054 | 40001465 | ||
BC100833.1 | - | 7, 8 | 1054 | 40001467 | ||
BC100834.1 | - | 7, 8 | 1002 | |||
BC100835.1 | - | 7, 8 | 1054 | 40001470 |