Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308154.1 | NP_001295083.1 | 2 | 235 | ||
NM_198686.2 | NP_941959.1 | 2 | 235 | |||
XM_017021277.1 | XP_016876766.1 | 3 | 495 | |||
GenBank mRNA | AB672631.2 | - | 2 | 204 | ||
AB672632.1 | - | 2 | 205 | |||
AB678452.1 | - | 2 | 235 | |||
AB678453.1 | - | 2 | 218 | |||
BC040679.2 | - | 2 | 158 | 4817835 | ||
BX248046.1 | - | 1 | 31 | |||
BX640825.1 | - | 2 | 118 | |||
CN352234.1 | - | 2 | 236 |