Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198581.2 | NP_940983.2 | 12 | 2825 | ||
XM_006712519.2 | XP_006712582.1 | 12 | 2832 | |||
GenBank mRNA | AB111887.1 | - | 11 | 2271 | ||
AK123404.1 | - | 2 | 442 | |||
AK127776.1 | - | 9 | 1894 | |||
AK131416.1 | - | 12 | 2811 | |||
BC155540.1 | - | 11 | 2271 | |||
BC167154.1 | - | 11 | 2271 |