Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP85L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042475.2 | NP_001035940.1 | 3 | 1197 | ||
NM_001178035.1 | NP_001171506.1 | 4 | 685 | |||
NM_206921.2 | NP_996804.2 | 3 | 1197 | |||
XM_005266970.1 | XP_005267027.1 | 3 | 717 | |||
XM_005266971.1 | XP_005267028.1 | 3 | 680 | |||
XM_006715475.3 | XP_006715538.1 | 4 | 893 | |||
XM_011535809.2 | XP_011534111.1 | 4 | 1788 | |||
XM_011535810.2 | XP_011534112.1 | 5 | 822 | |||
XM_011535811.1 | XP_011534113.1 | 3 | 583 | |||
XM_017010846.1 | XP_016866335.1 | 5 | 821 | |||
XM_017010847.1 | XP_016866336.1 | 3 | 1012 | |||
GenBank mRNA | AK302591.1 | - | 3 | 680 | ||
AY313778.1 | - | 3 | 653 | |||
BC036553.1 | - | 3 | 1234 | |||
BC048977.1 | - | 3 | 1197 | |||
BC052778.1 | - | 3 | 1197 | |||
BC110835.1 | - | 5 | 789 | |||
BC126140.1 | - | 2 | 493 | |||
BC126142.1 | - | 2 | 493 |